Merck
  • Alterations of Amphetamine Reward by Prior Nicotine and Alcohol Treatment: The Role of Age and Dopamine.
Alterations of Amphetamine Reward by Prior Nicotine and Alcohol Treatment: The Role of Age and Dopamine.
Brain sciences (2021-04-04)
Andrea Stojakovic, Syed Muzzammil Ahmad, Kabirullah Lutfy
ABSTRACT
Evidence suggests that nicotine and alcohol can each serve as a gateway drug. We determined whether prior nicotine and alcohol treatment would alter amphetamine reward. Also, we examined whether age and dopaminergic neurotransmission are important in this regard. Male and female adolescent and adult C57BL/6J mice were tested for baseline place preference. Mice then received six conditioning with saline/nicotine (0.25 mg/kg) twice daily, followed by six conditioning with saline/ethanol (2 g/kg). Control mice were conditioned with saline/saline throughout. Finally, mice were conditioned with amphetamine (3 mg/kg), once in the nicotine-alcohol-paired chamber, and tested for place preference 24 h later. The following day, mice were challenged with amphetamine (1 mg/kg) and tested for place preference under a drugged state. Mice were then immediately euthanized, their brain removed, and nucleus accumbens isolated and processed for the level of dopamine receptors and transporter and glutamate receptors. We observed a greater amphetamine-induced place preference in naïve adolescents than adult mice with no change in state-dependent place preference between the two age groups. In contrast, amphetamine induced a significant place preference in adult but not adolescent mice with prior nicotine-alcohol exposure under the drug-free state. The preference was significantly greater in adults than adolescents under the drugged state. The enhanced response was associated with higher dopamine-transporter and D1 but reduced D2 receptors' expression in adult rather than adolescent mice, with no changes in glutamate receptors levels. These results suggest that prior nicotine and alcohol treatment differentially alters amphetamine reward in adult and adolescent mice. Alterations in dopaminergic neurotransmission may be involved in this phenotype.
Sigma-Aldrich
Anti-Dopamine D(1A) Receptor Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Dopamine Transporter; 100 µg Antibody, from rabbit
Sigma-Aldrich
Anti-Dopamine D2 Receptor Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®