Merck
  • Dynamics and competition of CRISPR-Cas9 ribonucleoproteins and AAV donor-mediated NHEJ, MMEJ and HDR editing.
Dynamics and competition of CRISPR-Cas9 ribonucleoproteins and AAV donor-mediated NHEJ, MMEJ and HDR editing.
Nucleic acids research (2021-01-06)
Ya-Wen Fu, Xin-Yue Dai, Wen-Tian Wang, Zhi-Xue Yang, Juan-Juan Zhao, Jian-Ping Zhang, Wei Wen, Feng Zhang, Kerby C Oberg, Lei Zhang, Tao Cheng, Xiao-Bing Zhang
ABSTRACT
Investigations of CRISPR gene knockout editing profiles have contributed to enhanced precision of editing outcomes. However, for homology-directed repair (HDR) in particular, the editing dynamics and patterns in clinically relevant cells, such as human iPSCs and primary T cells, are poorly understood. Here, we explore the editing dynamics and DNA repair profiles after the delivery of Cas9-guide RNA ribonucleoprotein (RNP) with or without the adeno-associated virus serotype 6 (AAV6) as HDR donors in four cell types. We show that editing profiles have distinct differences among cell lines. We also reveal the kinetics of HDR mediated by the AAV6 donor template. Quantification of T50 (time to reach half of the maximum editing frequency) indicates that short indels (especially +A/T) occur faster than longer (>2 bp) deletions, while the kinetics of HDR falls between NHEJ (non-homologous end-joining) and MMEJ (microhomology-mediated end-joining). As such, AAV6-mediated HDR effectively outcompetes the longer MMEJ-mediated deletions but not NHEJ-mediated indels. Notably, a combination of small molecular compounds M3814 and Trichostatin A (TSA), which potently inhibits predominant NHEJ repairs, leads to a 3-fold increase in HDR efficiency.
Sigma-Aldrich
RAD51-Stimulatory Compound-1, RS-1, A cell-permeable sulfonamido-benzamide-based allosteric regulator that stimulates DNA binding and recombinase activities of hRAD51 by locking hRAD51 in an active conformation without affecting its active site ATP hydrolysis.
Pricing and availability is not currently available.
Sigma-Aldrich
RAD51 Inhibitor B02, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Cyclosporin H, ≥97% (HPLC)
Pricing and availability is not currently available.