Merck
  • Pyronaridine tetraphosphate efficacy against Ebola virus infection in guinea pig.
Pyronaridine tetraphosphate efficacy against Ebola virus infection in guinea pig.
Antiviral research (2020-07-20)
Thomas R Lane, Christopher Massey, Jason E Comer, Alexander N Freiberg, Huanying Zhou, Julie Dyall, Michael R Holbrook, Manu Anantpadma, Robert A Davey, Peter B Madrid, Sean Ekins
ABSTRACT
The recent outbreaks of the Ebola virus (EBOV) in Africa have brought global visibility to the shortage of available therapeutic options to treat patients infected with this or closely related viruses. We have recently computationally identified three molecules which have all demonstrated statistically significant efficacy in the mouse model of infection with mouse adapted Ebola virus (ma-EBOV). One of these molecules is the antimalarial pyronaridine tetraphosphate (IC50 range of 0.82-1.30 μM against three strains of EBOV and IC50 range of 1.01-2.72 μM against two strains of Marburg virus (MARV)) which is an approved drug in the European Union and used in combination with artesunate. To date, no small molecule drugs have shown statistically significant efficacy in the guinea pig model of EBOV infection. Pharmacokinetics and range-finding studies in guinea pigs directed us to a single 300 mg/kg or 600 mg/kg oral dose of pyronaridine 1hr after infection. Pyronaridine resulted in statistically significant survival of 40% at 300 mg/kg and protected from a lethal challenge with EBOV. In comparison, oral favipiravir (300 mg/kg dosed once a day) had 43.5% survival. All animals in the vehicle treatment group succumbed to disease by study day 12 (100% mortality). The in vitro metabolism and metabolite identification of pyronaridine and another of our EBOV active molecules, tilorone, suggested significant species differences which may account for the efficacy or lack thereof, respectively in guinea pig. In summary, our studies with pyronaridine demonstrates its utility for repurposing as an antiviral against EBOV and MARV.
Sigma-Aldrich
Triton X-100, Molecular Biology
Sigma-Aldrich
Triton X-100, laboratory grade