Merck
  • Topical application of Zanthoxylum piperitum extract improves lateral canthal rhytides by inhibiting muscle contractions.
Topical application of Zanthoxylum piperitum extract improves lateral canthal rhytides by inhibiting muscle contractions.
Scientific reports (2020-12-11)
Wooseon Hwang, Daehyun Kim, Oh Sun Kwon, Yun-Sun Kim, Byungjun Ahn, Nae-Gyu Kang
ABSTRACT
Facial wrinkles are the predominant phenotypes of skin aging. To date, one of the most effective ways to improve wrinkles is botulinum toxin type A (BoNT/A) injection, which inhibits muscle contractions by reducing acetylcholine release from neurons. However, since BoNT/A is a hazardous neurotoxin, the injection can only be performed by medical doctors and the procedure is only possible through invasive injection, causing inconveniences such as pain. To overcome these inconveniences, we tried to find a way to reduce wrinkles non-invasively via mechanisms similar to BoNT/A. We first designed in vitro assays to test BoNT/A-like muscle contraction inhibition in two different model systems. By using the assays, we identified Zanthoxylum piperitum (Z. piperitum) fruit extract as a BoNT-like reagent (27.7% decrease of muscle contraction rates by 1000 ppm of Z. piperitum extract treatment). Next, we determined mechanisms of how Z. piperitum extract decreases muscle contraction rates and found that the extract treatment inhibits electrical signal transduction in neurons. We also showed that among known components of Z. piperitum extract, quercitrin is responsible for muscle contraction inhibition. We further identified that Z. piperitum extract has synergistic effects with acetyl hexapeptide-8 and BoNT/A light chain, which are well-known BoNT-like peptides. Finally, we showed that topical treatment of the Z. piperitum extract indeed decreases facial wrinkles and treatment of Z. piperitum extract with acetyl hexapeptide-8 has a tendency to improve wrinkles synergistically (14.5% improvement on average). The synergistic effect of the combination is expected to improve wrinkles effectively by implementing the BoNT/A mechanisms in a non-invasive way.
Sigma-Aldrich
Ac-EEMQRR-NH2 trifluoroacetate salt, ≥95% (HPLC)
Pricing and availability is not currently available.