Merck
  • Annexin A1-dependent tethering promotes extracellular vesicle aggregation revealed with single-extracellular vesicle analysis.
Annexin A1-dependent tethering promotes extracellular vesicle aggregation revealed with single-extracellular vesicle analysis.
Science advances (2020-09-18)
Maximillian A Rogers, Fabrizio Buffolo, Florian Schlotter, Samantha K Atkins, Lang H Lee, Arda Halu, Mark C Blaser, Elena Tsolaki, Hideyuki Higashi, Kristin Luther, George Daaboul, Carlijn V C Bouten, Simon C Body, Sasha A Singh, Sergio Bertazzo, Peter Libby, Masanori Aikawa, Elena Aikawa
ABSTRACT
Extracellular vesicles (EVs) including plasma membrane-derived microvesicles and endosomal-derived exosomes aggregate by unknown mechanisms, forming microcalcifications that promote cardiovascular disease, the leading cause of death worldwide. Here, we show a framework for assessing cell-independent EV mechanisms in disease by suggesting that annexin A1 (ANXA1)-dependent tethering induces EV aggregation and microcalcification. We present single-EV microarray, a method to distinguish microvesicles from exosomes and assess heterogeneity at a single-EV level. Single-EV microarray and proteomics revealed increased ANXA1 primarily on aggregating and calcifying microvesicles. ANXA1 vesicle aggregation was suppressed by calcium chelation, altering pH, or ANXA1 neutralizing antibody. ANXA1 knockdown attenuated EV aggregation and microcalcification formation in human cardiovascular cells and acellular three-dimensional collagen hydrogels. Our findings explain why microcalcifications are more prone to form in vulnerable regions of plaque, regulating critical cardiovascular pathology, and likely extend to other EV-associated diseases, including autoimmune and neurodegenerative diseases and cancer.
Product Description
Supelco
L-Histidine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Pricing and availability is not currently available.