Merck
Water Electrolysis in Saturated Phosphate Buffer at Neutral pH.
ChemSusChem (2020-09-03)
Takahiro Naito, Tatsuya Shinagawa, Takeshi Nishimoto, Kazuhiro Takanabe
ABSTRACT
Hydrogen production from renewable energy and ubiquitous water has a potential to achieve sustainability, although current water electrolyzers cannot compete economically with the fossil fuel-based technology. Here, we evaluate water electrolysis at pH 7 that is milder than acidic and alkaline pH counterparts and may overcome this issue. The physicochemical properties of concentrated buffer electrolytes were assessed at various temperatures and molalities for quantitative determination of losses associated with mass-transport during the water electrolysis. Subsequently, in saturated K-phosphate solutions at 80 °C and 100 °C that were found to be optimal to minimize the losses originating from mass-transport at the neutral pH, the water electrolysis performance over model electrodes of IrOx and Pt as an anode and a cathode, respectively, was reasonably comparable with those of the extreme pH. Remarkably, this concentrated buffer solution also achieved enhanced stability, adding another merit of this electrolyte for water electrolysis.
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Nitric acid, SAJ first grade, 69-70%, density: 1.42