Merck
  • Water-Borne Isocyanate-Free Polyurethane Hydrogels with Adaptable Functionality and Behavior.
Water-Borne Isocyanate-Free Polyurethane Hydrogels with Adaptable Functionality and Behavior.
Macromolecular rapid communications (2020-10-14)
Maxime Bourguignon, Jean-Michel Thomassin, Bruno Grignard, Bénédicte Vertruyen, Christophe Detrembleur
ABSTRACT
Polyurethane hydrogels are attractive materials finding multiple applications in various sectors of prime importance; however, they are still prepared by the toxic isocyanate chemistry. Herein the facile and direct preparation in water at room temperature of a large palette of anionic, cationic, or neutral polyurethane hydrogels by a non-isocyanate route from readily available diamines and new hydrosoluble polymers bearing cyclic carbonates is reported. The latter are synthesized by free radical polymerization of glycerin carbonated methacrylate with water-soluble comonomers. The hydrogel formation is studied at different pH and its influence on the gel time and storage modulus is investigated. Reinforced hydrogels are also constructed by adding CaCl2 to the formulation that in-situ generates CaCO3 particles. Thermoresponsive hydrogels are also prepared from new thermoresponsive cyclic carbonate bearing polymers. This work demonstrates that a multitude of non-isocyanate polyurethane hydrogels are easily accessible under mild conditions without any catalyst, opening new perspectives in the field.
Sigma-Aldrich
Nonafluoro-tert-butyl alcohol, 99%
Sigma-Aldrich
2-Acrylamido-2-methyl-1-propanesulfonic acid, 99%