Merck
  • PbS/CdS Quantum Dot Room-Temperature Single-Emitter Spectroscopy Reaches the Telecom O and S Bands via an Engineered Stability.
PbS/CdS Quantum Dot Room-Temperature Single-Emitter Spectroscopy Reaches the Telecom O and S Bands via an Engineered Stability.
ACS nano (2021-01-01)
Sachidananda Krishnamurthy, Ajay Singh, Zhongjian Hu, Anastasia V Blake, Younghee Kim, Amita Singh, Ekaterina A Dolgopolova, Darrick J Williams, Andrei Piryatinski, Anton V Malko, Han Htoon, Milan Sykora, Jennifer A Hollingsworth
ABSTRACT
We synthesized PbS/CdS core/shell quantum dots (QDs) to have functional single-emitter properties for room-temperature, solid-state operation in the telecom O and S bands. Two shell-growth methods-cation exchange and successive ionic layer adsorption and reaction (SILAR)-were employed to prepare QD heterostructures with shells of 2-16 monolayers. PbS/CdS QDs were sufficiently bright and stable to resolve photoluminescence (PL) spectra representing both bands from single nanocrystals using standard detection methods, and for a QD emitting in the O-band a second-order correlation function showed strong photon antibunching, important steps toward demonstrating the utility of lead chalcogenide QDs as single-photon emitters (SPEs). Irrespective of type, few telecom-SPEs exist that are capable of such room-temperature operation. Access to single-QD spectra enabled a direct assessment of spectral line width, which was ∼70-90 meV compared to much broader ensemble spectra (∼300 meV). We show inhomogeneous broadening results from dispersity in PbS core sizes that increases dramatically with extended cation exchange. Quantum yields (QYs) are negatively impacted at thick shells (>6 monolayers) and, especially, by SILAR-growth conditions. Time-resolved PL measurements revealed that, with SILAR, initially single-exponential PL-decays transition to biexponential, with opening of nonradiative carrier-recombination channels. Radiative decay times are, overall, longer for core/shell QDs compared to PbS cores, which we demonstrate can be partially attributed to some core/shell sizes occupying a quasi-type II electron-hole localization regime. Finally, we demonstrate that shell engineering and the use of lower laser-excitation powers can afford significantly suppressed blinking and photobleaching. However, dependence on shell thickness comes at a cost of less-than-optimal brightness, with implications for both materials and experimental design.
Sigma-Aldrich
Cadmium oxide, ≥99.99% trace metals basis
Pricing and availability is not currently available.