Merck
  • Simultaneous quantification method for 5-FU, uracil, and tegafur using UPLC-MS/MS and clinical application in monitoring UFT/LV combination therapy after hepatectomy.
Simultaneous quantification method for 5-FU, uracil, and tegafur using UPLC-MS/MS and clinical application in monitoring UFT/LV combination therapy after hepatectomy.
Scientific reports (2021-02-06)
Ken Shiraiwa, Yosuke Suzuki, Hiroki Uchida, Yukio Iwashita, Ryota Tanaka, Motoshi Iwao, Kazuhiro Tada, Teijiro Hirashita, Takashi Masuda, Yuichi Endo, Masafumi Inomata, Hiroki Itoh
ABSTRACT
Combination therapy of tegafur/uracil (UFT) and leucovorin (LV) is widely used to treat colorectal cancers. Although this therapy has a significant therapeutic effect, severe adverse effects occur frequently. Therapeutic drug monitoring (TDM) may help to prevent adverse effects. A useful assay that can quantitate plasma levels of 5-FU, uracil, and tegafur simultaneously for TDM has been desired, but such a method is not currently available. In this study, we aimed to develop a sensitive method for simultaneous quantification of 5-FU, uracil, and tegafur in human plasma using ultra-performance liquid chromatography coupled to tandem mass spectrometry (UPLC-MS/MS). After preparing plasma samples by protein precipitation and liquid extraction, 5-FU, uracil, and tegafur were analyzed by UPLC-MS/MS in negative electrospray ionization mode. Validation was performed according to US Food and Drugs Administration guidance. The calibration curves were linear over concentration ranges of 2-500 ng/mL for 5-FU, 20-5000 ng/mL for uracil, and 200-50,000 ng/mL for tegafur. The corresponding average recovery rates were 79.9, 80.9, and 87.8%. The method provides accuracy within 11.6% and precision below 13.3% for all three analytes. Matrix effects of 5-FU, uracil, and tegafur were higher than 43.5, 84.9, and 100.2%, respectively. This assay was successfully applied to assess the time courses of plasma 5-FU, uracil, and tegafur concentrations in two patients with colorectal liver metastasis who received UFT/LV therapy after hepatectomy. In conclusion, we succeeded to develop a sensitive and robust UPLC-MS/MS method for simultaneous quantification of 5-FU, uracil, and tegafur in human plasma. This method is potentially useful for TDM in patients receiving UFT/LV combination therapy.
Sigma-Aldrich
Uracil-15N2, 98 atom % 15N
Pricing and availability is not currently available.