Merck
  • A Crucial Role of ACBD3 Required for Coxsackievirus Infection in Animal Model Developed by AAV-Mediated CRISPR Genome Editing Technique.
A Crucial Role of ACBD3 Required for Coxsackievirus Infection in Animal Model Developed by AAV-Mediated CRISPR Genome Editing Technique.
Viruses (2021-02-07)
Hye Jin Shin, Keun Bon Ku, Soojin Kim, Heon Seok Kim, Yeon-Soo Kim, Bum-Tae Kim, Seong-Jun Kim, Chonsaeng Kim
ABSTRACT
Genetic screens using CRISPR/Cas9 have been exploited to discover host-virus interactions. These screens have identified viral dependencies on host proteins during their life cycle and potential antiviral strategies. The acyl-CoA binding domain containing 3 (ACBD3) was identified as an essential host factor for the Coxsackievirus B3 (CVB3) infection. Other groups have also investigated the role of ACBD3 as a host factor for diverse enteroviruses in cultured cells. However, it has not been tested if ACBD3 is required in the animal model of CVB3 infection. Owing to embryonic lethality, conventional knockout mice were not available for in vivo study. As an alternative approach, we used adeno-associated virus (AAV)-mediated CRISPR genome editing to generate mice that lacked ACBD3 within the pancreas, the major target organ for CVB3. Delivery of sgRNAs using self-complementary (sc) AAV8 efficiently induced a loss-of-function mutation in the pancreas of the Cas9 knock-in mice. Loss of ACBD3 in the pancreas resulted in a 100-fold reduction in the CVB3 titer within the pancreas and a noticeable reduction in viral protein expression. These results indicate a crucial function of ACBD3 in CVB3 infection in vivo. AAV-mediated CRISPR genome editing may be applicable to many in vivo studies on the virus-host interaction and identify a novel target for antiviral therapeutics.
Sigma-Aldrich
Anti-ACBD3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.
Nunc® Cell Factory, Nunclon® Δ surface, 10-chamber CF, 6320 cm2, 2000 mL, 6/cs
Pricing and availability is not currently available.