Merck
  • Diosgenin, a plant-derived sapogenin, enhances regulatory T-cell immunity in the intestine of mice with food allergy.
Diosgenin, a plant-derived sapogenin, enhances regulatory T-cell immunity in the intestine of mice with food allergy.
Journal of natural products (2010-05-15)
Chung-Hsiung Huang, Der-Zen Liu, Tong-Rong Jan
ABSTRACT
It was hypothesized that the suppressive effect of diosgenin (1) on the intestinal T helper (Th)2 responses is associated with an enhancement of the regulatory T-cell immunity. Ovalbumin (OVA)-sensitized BALB/c mice were gavaged daily with 1 and received repeatedly oral OVA challenges to induce intestinal allergic responses. The expression of Th2- and Treg-related cytokines and transcription factors was examined by immunohistochemical staining and RT-PCR. Administration of 1 markedly attenuated the intestinal expression of interleukin (IL)-4 and GATA3. In addition, administration of 1 reversed the diminished density of intestinal Foxp3(+) cells induced by OVA oral challenges and enhanced the expression of IL-10 by Foxp3(+) cells markedly. These results suggest that the suppressive effect of 1 on allergen-induced intestinal Th2 responses is closely associated with an up-regulation of the regulatory T-cell immunity in the inflammatory site.
Sigma-Aldrich
Diosgenin, ≥93%