Merck
  • Characterizing the Dynamic Taste and Retro-Nasal Aroma Properties of Oral Nutritional Supplements Using Temporal Dominance of Sensation and Temporal Check-All-That-Apply Methods.
Characterizing the Dynamic Taste and Retro-Nasal Aroma Properties of Oral Nutritional Supplements Using Temporal Dominance of Sensation and Temporal Check-All-That-Apply Methods.
Foods (Basel, Switzerland) (2020-10-18)
Thomas Delompré, Louis Lenoir, Christophe Martin, Loïc Briand, Christian Salles
ABSTRACT
Orally Disintegrating Tablets (ODTs) are used to restore the nutritional status of people suffering from swallowing pathologies such as dysphagia. ODTs are consumed without water intake and are easily swallowed. The main active compounds of ODTs are vitamins and minerals. These nutrients can have a bad taste or aftertaste, which can be masked by sweetening or flavoring. To ensure the effectiveness of masking strategies and to prescribe a product to patients with acceptable sensory qualities, it is essential to perform a precise and complete sensory characterization of these ODTs. Temporal dominance of sensations (TDS) and temporal check-all-that-apply (TCATA) methods were chosen to characterize the temporal sensory perceptions during the consumption of four ODTs varying in galenic forms and flavoring because of their particularly acute temporality in flavor perception. The ODTs presented common and individual sensory properties, mainly related to their galenic form and to the nature of the active ingredients. The use of a nose-clip to stop retro-nasal airflow showed that flavoring had a minimal impact on the sensory taste qualities of the ODTs. A comparison between the TDS and TCATA results indicates that these tests are capable of providing complementary information on the dynamic sensory qualities of the products studied. Although results were generally similar for both methods, TDS showed a greater number of differences in sensory taste attribute, whereas TCATA was more discriminating. These methods allowed for a better understanding of the evolution of sensory perceptions of these ODTs during their consumption, which could help to optimize masking strategies and develop new products with acceptable sensory properties.
Product Description
Supelco
Ferrous Sulfate, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.