Merck
  • Optical Response Characteristics of Single-Walled Carbon Nanotube Chirality Exposed to Oxidants with Different Oxidizing Power.
Optical Response Characteristics of Single-Walled Carbon Nanotube Chirality Exposed to Oxidants with Different Oxidizing Power.
Molecules (Basel, Switzerland) (2021-03-07)
Yuji Matsukawa, Kazuo Umemura
ABSTRACT
Semiconductor single-walled carbon nanotubes (SWNTs) have unique characteristics owing to differences in the three-dimensional structure (chirality) expressed by the chiral index (n,m), and many studies on the redox characteristics of chirality have been reported. In this study, we investigated the relationship between the chirality of SWNTs and the oxidizing power of oxidants by measuring the near-infrared (NIR) absorption spectra of two double-stranded DNA-SWNT complexes with the addition of three oxidants with different oxidizing powers. A dispersion was prepared by mixing 0.5 mg of SWNT powder with 1 mg/mL of DNA solution. Different concentrations of hydrogen peroxide (H2O2), potassium hexachloroidylate (IV) (K2IrCl6), or potassium permanganate (KMnO4) were added to the dispersion to induce oxidation. Thereafter, a catechin solution was added to observe if the absorbance of the oxidized dispersion was restored by the reducing action of the catechin. We found that the difference in the oxidizing power had a significant effect on the detection sensitivity of the chiralities of the SWNTs. Furthermore, we revealed a detectable range of oxidants with different oxidizing powers for each chirality.
Sigma-Aldrich
Carbon nanotube, single-walled, (6,5) chirality, ≥95% carbon basis (≥95% as carbon nanotubes), 0.78 nm average diameter
Sigma-Aldrich
Fluorescein (free acid), Dye content 95 %
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from salmon testes