Merck
  • Synthesis and anti-leishmanial activity of heterocyclic betulin derivatives.
Synthesis and anti-leishmanial activity of heterocyclic betulin derivatives.
Bioorganic & medicinal chemistry (2010-02-02)
Sami Alakurtti, Tuomo Heiska, Alexandros Kiriazis, Nina Sacerdoti-Sierra, Charles L Jaffe, Jari Yli-Kauhaluoma
ABSTRACT
Betulin, a naturally occurring abundant triterpene is converted in four steps to 3,28-di-O-acetyllupa-12,18-diene. When various 4-substituted urazoles were oxidized to the corresponding urazines with iodobenzene diacetate in the presence of 3,28-di-O-acetyllupa-12,18-diene, new heterocyclic betulin derivatives were produced. These betulin derivatives were examined in a microplate assay at 50 microM for their ability to inhibit the growth of Leishmania donovani axenic amastigotes, a species that causes the fatal visceral leishmaniasis. GI(50) (concentration for 50% growth inhibition) values of the most effective compounds were determined and their cytotoxicity on the human macrophage cell line THP-1 evaluated. The anti-leishmanial activity on L. donovani amastigotes growing in macrophages was also examined. The heterocycloadduct between 3,28-di-O-acetyllupa-12,18-diene and 4-methylurazine was the most effective derivative with an GI(50)=8.9 microM against L. donovani amastigotes.
Sigma-Aldrich
Betulinic acid, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Betulinic acid, technical grade, 90%
Pricing and availability is not currently available.