Merck
  • Calibration of Quartz-Enhanced Photoacoustic Sensors for Real-Life Adaptation.
Calibration of Quartz-Enhanced Photoacoustic Sensors for Real-Life Adaptation.
Molecules (Basel, Switzerland) (2021-01-29)
Jesper B Christensen, David Balslev-Harder, Lars Nielsen, Jan C Petersen, Mikael Lassen
ABSTRACT
We report on the use of quartz-enhanced photoacoustic spectroscopy for continuous carbon-dioxide measurements in humid air over a period of six days. The presence of water molecules alters the relaxation rate of the target molecules and thus the amplitude of the photoacoustic signal. Prior to the measurements, the photoacoustic sensor system was pre-calibrated using CO2 mole fractions in the range of 0-10-3 (0-1000 ppm) and at different relative humidities between 0% and 45%, while assuming a model hypothesis that allowed the photoacoustic signal to be perturbed linearly by H2O content. This calibration technique was compared against an alternative learning-based method, where sensor data from the first two days of the six-day period were used for self-calibration. A commercial non-dispersive infrared sensor was used as a CO2 reference sensor and provided the benchmark for the two calibration procedures. In our case, the self-calibrated method proved to be both more accurate and precise.
Sigma-Aldrich
Lithium niobate, 99.9% trace metals basis
Pricing and availability is not currently available.