Merck
  • Screening of antimicrobial synergism between phenolic acids derivatives and UV-A light radiation.
Screening of antimicrobial synergism between phenolic acids derivatives and UV-A light radiation.
Journal of photochemistry and photobiology. B, Biology (2020-11-27)
Erick F de Oliveira, Xu Yang, Nikeshala Basnayake, Cuong Nguyen Huu, Luxin Wang, Rohan Tikekar, Nitin Nitin
ABSTRACT
The objective of this study was to investigate synergistic antibacterial activity based on a combination of UV-A light and three classes of food grade compounds: benzoic acid derivatives, cinnamic acid derivatives, and gallates. By using Escherichia coli O157:H7 as the model strain, it was observed that three cinnamic acid derivatives (ferulic acid, coumaric acid, and caffeic acid) and one benzoic acid derivative (2,5-dihydroxybenzoic acid) presented strong synergistic antibacterial activity with UV-A light radiation, where 1 mM levels of these compounds plus with 15 min of UV-A light (total light dose of 6.1 cm-2) led to more than 7-log CFU mL-1 of bacterial inactivation. In contrast, synergistic antibacterial activity between UV-A light and most benzoic acid derivatives (benzoic acid, gallic acid, vanillic acid, and 2,5-dimethoxybenzoic acid) were only observed after higher concentrations of these compounds were applied (10 mM). Lastly, from the three gallates tested (methyl gallate, ethyl gallate, and propyl gallate), only propyl gallate showed strong antibacterial synergism with UV-A light, where 10 mM of propyl gallate plus 15 min of UV-A light led to approximately 6.5-log of bacterial reduction. Presence of antioxidant compounds mitigated the light-mediated antibacterial activity of gallic acid, 2,5-dihydroxybenzoic acid, and propyl gallate. Similarly, the light-mediated antibacterial activity of these compounds was significantly (P < 0.05) reduced against metabolic-inhibited bacterial cells (sodium azide pretreatment). On the other hand, the antibacterial synergism between ferulic acid and UV-A light was not affected by the presence of antioxidants or the metabolic state of the bacterial cells. Due to the increasing concerns of antimicrobial resistant (AMR) pathogens, the study also investigated the proposed synergistic treatment on AMR Salmonella. Combinations of 1 mM of ferulic acid or 1 mM of 2,5-dihydroxybenzoic acid with UV-A light radiation was able to inactivate more than 6-log of a multi-drug resistant Salmonella Typhimurium strain.