Merck
  • A role for 4-hydroxy-2-nonenal in premature placental senescence in preeclampsia and intrauterine growth restriction.
A role for 4-hydroxy-2-nonenal in premature placental senescence in preeclampsia and intrauterine growth restriction.
Free radical biology & medicine (2021-01-16)
Oriane Tasta, Audrey Swiader, Marie-Hélène Grazide, Myriam Rouahi, Olivier Parant, Christophe Vayssière, Emmanuel Bujold, Robert Salvayre, Paul Guerby, Anne Negre-Salvayre
ABSTRACT
Premature placental senescence is a hallmark of pregnancy-related disorders such as intrauterine growth restriction (IUGR) and preeclampsia (PE), two major cause of maternal and neonatal morbidity and mortality. Oxidative stress and lipid peroxidation are involved in the pathogenesis of PE and IUGR, and may play a role in placental aging. In this study, we investigated whether 4-hydroxy-2-nonenal (HNE), a lipid peroxidation-derived aldehyde present in preeclamptic placentas, may contribute to premature senescence in placenta-related complications. Placentas from PE-affected women, exhibited several senescence patterns, such as an increased expression of phosphorylated (serine-139) histone γH2AX, a sensitive marker of double-stranded DNA breaks, the presence of lipofuscin granules, and an accumulation of high molecular weight cross-linked and ubiquitinated proteins. PE placentas showed an accumulation of acetylated proteins consistent with the presence of HNE-adducts on sirtuin 1 (SIRT1). Likewise, oxidative stress and senescence markers together with SIRT1 modification by HNE, were observed in murine placentas from mice treated with lipopolysaccharide during gestation and used as models of IUGR. The addition of HNE and ONE (4-oxo-2-nonenal), to cultured HTR-8/SVneo human trophoblasts activated the senescence-associated- β-galactosidase, and generated an accumulation of acetylated proteins, consistent with a modification of SIRT1 by HNE. Altogether, these data emphasize the role of HNE and lipid peroxidation-derived aldehydes in premature placental senescence in PE and IUGR, and more generally in pathological pregnancies.