Merck
  • OXA-143, a novel carbapenem-hydrolyzing class D beta-lactamase in Acinetobacter baumannii.
OXA-143, a novel carbapenem-hydrolyzing class D beta-lactamase in Acinetobacter baumannii.
Antimicrobial agents and chemotherapy (2009-09-23)
Paul G Higgins, Laurent Poirel, Marlene Lehmann, Patrice Nordmann, Harald Seifert
ABSTRACT
A carbapenem-resistant Acinetobacter baumannii strain was isolated in Brazil in 2004 in which no known carbapenemase gene was detected by PCR. Cloning experiments, followed by expression in Escherichia coli, gave an E. coli recombinant strain expressing a novel carbapenem-hydrolyzing class D beta-lactamase (CHDL). OXA-143 showed 88% amino acid sequence identity with OXA-40, 63% identity with OXA-23, and 52% identity with OXA-58. It hydrolyzed penicillins, oxacillin, meropenem, and imipenem but not expanded-spectrum cephalosporins. The bla(OXA-143) gene was located on a ca. 30-kb plasmid. After transformation into reference strain A. baumannii ATCC 19606, it conferred resistance to carbapenems. Analysis of the genetic environment of bla(OXA-143) revealed that it was associated with neither insertion sequences nor integron structures. However, it was bracketed by similar replicase-encoding genes at both ends, suggesting acquisition through a homologous recombination process. This study identified a novel class D beta-lactamase involved in carbapenem resistance in A. baumannii. This enzyme is the first member of a novel subgroup of CHDLs whose prevalence remains to be determined.
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Pricing and availability is not currently available.
Sigma-Aldrich
Amoxicillin, 95.0-102.0% anhydrous basis
Pricing and availability is not currently available.