Merck
  • Genome-wide screens identify Toxoplasma gondii determinants of parasite fitness in IFNγ-activated murine macrophages.
Genome-wide screens identify Toxoplasma gondii determinants of parasite fitness in IFNγ-activated murine macrophages.
Nature communications (2020-10-18)
Yifan Wang, Lamba Omar Sangaré, Tatiana C Paredes-Santos, Musa A Hassan, Shruthi Krishnamurthy, Anna M Furuta, Benedikt M Markus, Sebastian Lourido, Jeroen P J Saeij
ABSTRACT
Macrophages play an essential role in the early immune response against Toxoplasma and are the cell type preferentially infected by the parasite in vivo. Interferon gamma (IFNγ) elicits a variety of anti-Toxoplasma activities in macrophages. Using a genome-wide CRISPR screen we identify 353 Toxoplasma genes that determine parasite fitness in naїve or IFNγ-activated murine macrophages, seven of which are further confirmed. We show that one of these genes encodes dense granule protein GRA45, which has a chaperone-like domain, is critical for correct localization of GRAs into the PVM and secretion of GRA effectors into the host cytoplasm. Parasites lacking GRA45 are more susceptible to IFNγ-mediated growth inhibition and have reduced virulence in mice. Together, we identify and characterize an important chaperone-like GRA in Toxoplasma and provide a resource for the community to further explore the function of Toxoplasma genes that determine fitness in IFNγ-activated macrophages.
Product Description
Supelco
Mycophenolic acid, analytical standard
Sigma-Aldrich
5-Fluoro-2′-deoxyuridine, thymidylate synthase inhibitor
Sigma-Aldrich
Xanthine sodium salt, suitable for cell culture, ≥99%
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)