Merck
  • Near infrared-light responsive WS2 microengines with high-performance electro- and photo-catalytic activities.
Near infrared-light responsive WS2 microengines with high-performance electro- and photo-catalytic activities.
Chemical science (2020-02-29)
Víctor de la Asunción-Nadal, Beatriz Jurado-Sánchez, Luis Vázquez, Alberto Escarpa
ABSTRACT
Tungsten disulfide (WS2)-based micromotors with enhanced electrochemical and photo-catalytic activities are synthesized using a greatly simplified electrochemical deposition protocol at room temperature involving exclusively tungstic acid and sulfate as metal and sulfur sources without further building chemistry. The WS2-based micromotors exhibit dual electrochemical and photo-catalytic behavior in the inner and outer layers, respectively, due to the combination of the unique properties of the sp2 hybridized WS2 outer layer with highly reactive WS2-induced inner catalytic layers, accounting for this material's exclusive enhanced performances. A rough inner Pt-Ni layer allows tailoring the micromotor propulsion, with a speed increase of up to 1.6 times after external control of the micromotor with a magnetic field due to enhanced fuel accessibility. Such a coupling of the attractive capabilities of WS2 with enhanced micromotor movement holds considerable promise to address the growing energy crisis and environmental pollution concerns.
Sigma-Aldrich
Chloroplatinic acid hydrate, 99.995% trace metals basis
Pricing and availability is not currently available.