Merck
  • Intersection of the ATF6 and XBP1 ER stress pathways in mouse islet cells.
Intersection of the ATF6 and XBP1 ER stress pathways in mouse islet cells.
The Journal of biological chemistry (2020-08-14)
Rohit B Sharma, Christine Darko, Laura C Alonso
ABSTRACT
Success or failure of pancreatic beta cell adaptation to ER stress is a determinant of diabetes susceptibility. The ATF6 and IRE1/XBP1 pathways are separate ER stress-response effectors important to beta cell health and function. ATF6α. and XBP1 direct overlapping transcriptional responses in some cell types. However, the signaling dynamics and interdependence of ATF6α and XBP1 in pancreatic beta cells have not been explored. To assess pathway-specific signal onset, we performed timed exposures of primary mouse islet cells to ER stressors and measured the early transcriptional response. Comparing the time course of induction of ATF6 and XBP1 targets suggested that the two pathways have similar response dynamics. The role of ATF6α in target induction was assessed by acute knockdown using islet cells from Atf6α flox/flox mice transduced with adenovirus expressing Cre recombinase. Surprisingly, given the mild impact of chronic deletion in mice, acute ATF6α knockdown markedly reduced ATF6-pathway target gene expression under both basal and stressed conditions. Intriguingly, although ATF6α knockdown did not alter Xbp1 splicing dynamics or intensity, it did reduce induction of XBP1 targets. Inhibition of Xbp1 splicing did not decrease induction of ATF6α targets. Taken together, these data suggest that the XBP1 and ATF6 pathways are simultaneously activated in islet cells in response to acute stress and that ATF6α is required for full activation of XBP1 targets, but XBP1 is not required for activation of ATF6α targets. These observations improve understanding of the ER stress transcriptional response in pancreatic islets.
Sigma-Aldrich
IRE1 Inhibitor III, 4μ8C, IRE1 Inhibitor III, CAS 14003-96-4, is a cell-permeable. Covalent inhibitor of IRE1 RNase activity (IC₅₀ = 550 and 45 nM, respectively, with 0 & 16 min preincubation in RNA cleavage assays).
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film