Merck
  • Phospholipase D functional ablation has a protective effect in an Alzheimer's disease Caenorhabditis elegans model.
Phospholipase D functional ablation has a protective effect in an Alzheimer's disease Caenorhabditis elegans model.
Scientific reports (2018-02-25)
Francisca Vaz Bravo, Jorge Da Silva, Robin Barry Chan, Gilbert Di Paolo, Andreia Teixeira-Castro, Tiago Gil Oliveira
ABSTRACT
Phospholipase D (PLD) is a key player in the modulation of multiple aspects of cell physiology and has been proposed as a therapeutic target for Alzheimer's disease (AD). Here, we characterize a PLD mutant, pld-1, using the Caenorhabditis elegans animal model. We show that pld-1 animals present decreased phosphatidic acid levels, that PLD is the only source of total PLD activity and that pld-1 animals are more sensitive to the acute effects of ethanol. We further show that PLD is not essential for survival or for the normal performance in a battery of behavioral tests. Interestingly, pld-1 animals present both increased size and lipid stores levels. While ablation of PLD has no important effect in worm behavior, its ablation in an AD-like model that overexpresses amyloid-beta (Aβ), markedly improves various phenotypes such as motor tasks, prevents susceptibility to a proconvulsivant drug, has a protective effect upon serotonin treatment and reverts the biometric changes in the Aβ animals, leading to the normalization of the worm body size. Overall, this work proposes the C. elegans model as a relevant tool to study the functions of PLD and further supports the notion that PLD has a significant role in neurodegeneration.