Merck
  • Hierarchical Action of Mulberry miR156 in the Vegetative Phase Transition.
Hierarchical Action of Mulberry miR156 in the Vegetative Phase Transition.
International journal of molecular sciences (2021-06-03)
Hongshun Li, Yiwei Luo, Bi Ma, Jianqiong Hu, Zhiyuan Lv, Wuqi Wei, Haiye Hao, Jianglian Yuan, Ningjia He
ABSTRACT
The vegetative phase transition is a prerequisite for flowering in angiosperm plants. Mulberry miR156 has been confirmed to be a crucial factor in the vegetative phase transition in Arabidopsis thaliana. The over-expression of miR156 in transgenic Populus × canadensis dramatically prolongs the juvenile phase. Here, we find that the expression of mno-miR156 decreases with age in all tissues in mulberry, which led us to study the hierarchical action of miR156 in mulberry. Utilizing degradome sequencing and dual-luciferase reporter assays, nine MnSPLs were shown to be directly regulated by miR156. The results of yeast one-hybrid and dual-luciferase reporter assays also revealed that six MnSPLs could recognize the promoter sequences of mno-miR172 and activate its expression. Our results demonstrate that mno-miR156 performs its role by repressing MnSPL/mno-miR172 pathway expression in mulberry. This work uncovered a miR156/SPLs/miR172 regulation pathway in the development of mulberry and fills a gap in our knowledge about the molecular mechanism of vegetative phase transition in perennial woody plants.
Product Description
Roche
DIG DNA Labeling and Detection Kit
Pricing and availability is not currently available.