Merck
Crown-capped imidacloprid: a novel design and insecticidal activity.
Bioorganic & medicinal chemistry letters (2009-05-05)
Shinzo Kagabu, Masaru Takagi, Ikuya Ohno, Tsuyoshi Mikawa, Toru Miyamoto
ABSTRACT
Imidacloprid (IMI) derivatives conjugated with benzo-15-crown-5 and benzo-18-crown-6 structures, applied for the first time to explore novel insecticidal molecule, elicited strong excitatory toxic signs to the house flies and stunningly exhibited three to five times higher insecticidal activity than that of the parent IMI, yet the two benzo-crown structures themselves had no effect.
Sigma-Aldrich
15-Crown-5, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Benzo-15-crown-5, 98%
Pricing and availability is not currently available.