Merck
  • Design, synthesis, and biological activity of isophthalic acid derivatives targeted to the C1 domain of protein kinase C.
Design, synthesis, and biological activity of isophthalic acid derivatives targeted to the C1 domain of protein kinase C.
Journal of medicinal chemistry (2009-05-15)
Gustav Boije af Gennäs, Virpi Talman, Olli Aitio, Elina Ekokoski, Moshe Finel, Raimo K Tuominen, Jari Yli-Kauhaluoma
ABSTRACT
Protein kinase C (PKC) is a widely studied molecular target for the treatment of cancer and other diseases. We have approached the issue of modifying PKC function by targeting the C1 domain in the regulatory region of the enzyme. Using the X-ray crystal structure of the PKC delta C1b domain, we have discovered conveniently synthesizable derivatives of dialkyl 5-(hydroxymethyl)isophthalate that can act as potential C1 domain ligands. Structure-activity studies confirmed that the important functional groups predicted by modeling were indispensable for binding to the C1 domain and that the modifications of these groups diminished binding. The most promising compounds were able to displace radiolabeled phorbol ester ([(3)H]PDBu) from PKC alpha and delta at K(i) values in the range of 200-900 nM. Furthermore, the active isophthalate derivatives could modify PKC activation in living cells either by inducing PKC-dependent ERK phosphorylation or by inhibiting phorbol-induced ERK phosphorylation. In conclusion, we report here, for the first time, that derivatives of isophthalic acid represent an attractive novel group of C1 domain ligands that can be used as research tools or further modified for potential drug development.
Sigma-Aldrich
Diethyl 5-(hydroxymethyl)isophthalate, 98%
Sigma-Aldrich
Isophthalic acid, 99%