Merck
  • Sex-Dependent Dyslipidemia and Neuro-Humoral Alterations Leading to Further Cardiovascular Risk in Juvenile Obesity.
Sex-Dependent Dyslipidemia and Neuro-Humoral Alterations Leading to Further Cardiovascular Risk in Juvenile Obesity.
Frontiers in nutrition (2021-03-02)
Estefania Simoes, Joanna Correia-Lima, Elie Leal de Barros Calfat, Thais Zélia Dos Santos Otani, Daniel Augusto Correa Vasques, Victor Henrique Oyamada Otani, Pamela Bertolazzi, Cristiane Kochi, Marilia Seelaender, Ricardo Riyoiti Uchida
ABSTRACT
Objective: Childhood obesity is a growing concern as the World Health Organization (WHO) states that ~10% of adolescents worldwide are overweight or obese. This condition is the reflex of energy imbalance between the calories consumed and those expended. Sex-related responses associated with dyslipidemia, hormonal alterations, and neuro-humoral disruptions in childhood obesity are the focus of the present investigation. Methods: Ninety-two Brazilian adolescents were enrolled and divided between obese and eutrophic groups. Obesity was assessed using body mass index Z-score according to age and weight. Anthropometrical analyses, blood pressure, blood lipids, metabolism-regulating hormones, and neuropeptides were carried out. Results: Systolic blood pressure was higher in female and male patients with obesity. Obese females presented alterations in lipid profile and an augment of cardiovascular disease prediction ratios TC/HDL, TG/HDL, LDL/HDL, and VLDL/HDL. The levels of leptin, GIP, and neuropeptide showed sex-dimorphism in obesity. The obese adolescents presented increased levels of circulating insulin, c-peptide, amylin, glucagon, and GLP-1. Correlation analysis showed significant linearity between body mass index, blood pressure, lipids, lipoproteins, hormones, and neuropeptides content. Conclusions: Our data support an existing link associating hypertension, dyslipidemia, and neuro-hormonal imbalance in childhood obesity. We also described a sex-dependent pattern in childhood obesity-associated dyslipidemia and blood pressure in female patients with obesity solely.
Product Description
Millipore
MILLIPLEX® Human Metabolic Hormone Magnetic Bead Panel - Metabolism Multiplex Assay, The Human Metabolic Hormone Panel, using the Luminex xMAP technology, enables the simultaneous analysis of 13 protein biomarkers in human serum, plasma and cell/tissue culture samples.