Merck
  • CYTOKINE PROFILE ELEVATIONS ON ADMISSION CAN DETERMINE RISKS OF SEVERE ACUTE PANCREATITIS IN CHILDREN.
CYTOKINE PROFILE ELEVATIONS ON ADMISSION CAN DETERMINE RISKS OF SEVERE ACUTE PANCREATITIS IN CHILDREN.
The Journal of pediatrics (2021-07-18)
Peter R Farrell, Elaina K Jones, Lindsey Hornung, Tyler Thompson, Jyoti Patel, Tom K Lin, Jaimie D Nathan, David S Vitale, Aida Habtezion, Maisam Abu-El-Haija
ABSTRACT
To utilize a Luminex platform to examine multiple cytokines simultaneously as well as clinical laboratory testing in order to identify markers that predict acute pancreatitis (AP) severity in the pediatric population on admission. Patients (<19 years) prospectively enrolled over a 4-year period in a single institution AP database were included in separate derivation and validation cohorts. Plasma samples were obtained within 48 hours of admission and stored for analysis. Samples from mild AP and SAP (moderately severe and severe combined) were analyzed using Luminex panels and C-Reactive Protein (CRP) testing. The derivation cohort examined 62 cytokines in 66 subject samples (20 control, 36 mild AP, 10 SAP) and identified interleukin 6 (IL-6) [P = .02] and monocyte chemotactic protein-1 (MCP-1) [p=0.02] as cytokines that were differentially expressed between mild and SAP. Our validation cohort analyzed 76 cytokines between 10 controls, 19 mild AP and 6 SAP subjects. IL-6 (p=0.02) and MCP-1 (p=0.007) were again found to differentiate mild AP from SAP. CRP values were obtained from 53 of the subjects, revealing a strong association between elevated CRP values and progression to severe disease (P<0.0001). This study identified and validated IL-6 and MCP-1 as predictors of SAP using 2 distinct cohorts, and showed that CRP elevation is a marker of progression to SAP. These biomarkers have not been extensively studied in the pediatric AP population. Our data allows for risk-stratification of AP patients, and represent novel insight into the immunologic response in SAP.
Product Description
Millipore
MILLIPLEX® Human Cytokine/Chemokine Magnetic Bead Panel II - Premixed 23 Plex - Space Saver (Bulk) Packaging, Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in human serum, plasma and cell culture samples.
Pricing and availability is not currently available.