- Dataset on growth factor levels and insulin use in patients with diabetes mellitus and incident breast cancer.
Dataset on growth factor levels and insulin use in patients with diabetes mellitus and incident breast cancer.
Data in brief (2017-02-28)
Zachary A P Wintrob, Jeffrey P Hammel, George K Nimako, Dan P Gaile, Alan Forrest, Alice C Ceacareanu
PMID28239632
ABSTRACT
Growth factor profiles could be influenced by the utilization of exogenous insulin. The data presented shows the relationship between pre-existing use of injectable insulin in women diagnosed with breast cancer and type 2 diabetes mellitus, the growth factor profiles at the time of breast cancer diagnosis, and subsequent cancer outcomes. A Pearson correlation analysis evaluating the relationship between growth factors stratified by of insulin use and controls is also provided.