Merck
  • Mechanism of the protective effect of intraperitoneally administered agonists for formyl peptide receptors against chemotherapy-induced alopecia.
Mechanism of the protective effect of intraperitoneally administered agonists for formyl peptide receptors against chemotherapy-induced alopecia.
Bioscience, biotechnology, and biochemistry (2007-05-09)
Takahiro Tsuruki, Kyoya Takahata, Masaaki Yoshikawa
ABSTRACT
Previously, we found that an intraperitoneally administered chemotactic peptide, N-formyl-Met-Leu-Phe (fMLP), and MMK-1, a selective agonist of formyl peptide receptor-like 1 (FPRL1) receptor, the low affinity subtype of the fMLP receptor, prevented the alopecia in neonatal rats induced by the anticancer agent etoposide. The anti-alopecia effect of fMLP was not inhibited at all by Boc-FLFLF, a selective antagonist of formylpeptide receptor (FPR), which is the high affinity subtype of the fMLP receptor, but it was partly inhibited by Trp-Arg-Trp-Trp-Trp-Trp-NH(2) (WRW(4)), an antagonist of FPRL1 receptor. On the other hand, the anti-alopecia effect of MMK-1 was completely abolished by WRW(4). The anti-alopecia effects of fMLP and MMK-1 were also inhibited by Lys-D-Pro-Thr (K(D)PT) and pyrrolidine dithiocarbamate, which are inhibitors of interleukin-1 (IL-1) and nuclear factor-kappaB (NF-kappaB) respectively. Hence, we suggest that the anti-alopecia mechanisms of intraperitoneally administered fMLP and MMK-1 include activation of NF-kappaB via IL-1 release downstream of the FPRL1 receptor homolog in rats.
Sigma-Aldrich
MMK-1 trifluoroacetate, ≥95% (HPLC)