Merck
  • ORY-1001, a Potent and Selective Covalent KDM1A Inhibitor, for the Treatment of Acute Leukemia.
ORY-1001, a Potent and Selective Covalent KDM1A Inhibitor, for the Treatment of Acute Leukemia.
Cancer cell (2018-03-06)
Tamara Maes, Cristina Mascaró, Iñigo Tirapu, Angels Estiarte, Filippo Ciceri, Serena Lunardi, Nathalie Guibourt, Alvaro Perdones, Michele M P Lufino, Tim C P Somervaille, Dan H Wiseman, Cihangir Duy, Ari Melnick, Christophe Willekens, Alberto Ortega, Marc Martinell, Nuria Valls, Guido Kurz, Matthew Fyfe, Julio Cesar Castro-Palomino, Carlos Buesa
ABSTRACT
The lysine-specific demethylase KDM1A is a key regulator of stem cell potential in acute myeloid leukemia (AML). ORY-1001 is a highly potent and selective KDM1A inhibitor that induces H3K4me2 accumulation on KDM1A target genes, blast differentiation, and reduction of leukemic stem cell capacity in AML. ORY-1001 exhibits potent synergy with standard-of-care drugs and selective epigenetic inhibitors, reduces growth of an AML xenograft model, and extends survival in a mouse PDX (patient-derived xenograft) model of T cell acute leukemia. Surrogate pharmacodynamic biomarkers developed based on expression changes in leukemia cell lines were translated to samples from patients treated with ORY-1001. ORY-1001 is a selective KDM1A inhibitor in clinical trials and is currently being evaluated in patients with leukemia and solid tumors.
Sigma-Aldrich
N-Methyl-N-propargyl-3-(2,4-dichlorophenoxy)propylamine hydrochloride, ≥97% (GC)
Pricing and availability is not currently available.