Merck
  • Tenascin-C Deficiency Is Associated With Reduced Bacterial Outgrowth During Klebsiella pneumoniae-Evoked Pneumosepsis in Mice.
Tenascin-C Deficiency Is Associated With Reduced Bacterial Outgrowth During Klebsiella pneumoniae-Evoked Pneumosepsis in Mice.
Frontiers in immunology (2021-03-30)
Mariska T Meijer, Alex F de Vos, Brendon P Scicluna, Joris J Roelofs, Chérine Abou Fayçal, Gertraud Orend, Fabrice Uhel, Tom van der Poll
ABSTRACT
Tenascin C (TNC) is an extracellular matrix glycoprotein that recently emerged as an immunomodulator. TNC-deficient (TNC-/-) mice were reported to have a reduced inflammatory response upon systemic administration of lipopolysaccharide, the toxic component of gram-negative bacteria. Here, we investigated the role of TNC during gram-negative pneumonia derived sepsis. TNC+/+ and TNC-/- mice were infected with Klebsiella pneumoniae via the airways and sacrificed 24 and 42 h thereafter for further analysis. Pulmonary TNC protein levels were elevated 42 h after infection in TNC+/+ mice and remained undetectable in TNC-/- mice. TNC-/- mice showed modestly lower bacterial loads in lungs and blood, and a somewhat reduced local-but not systemic-inflammatory response. Moreover, TNC-/- and TNC+/+ mice did not differ with regard to neutrophil recruitment, lung pathology or plasma markers of distal organ injury. These results suggest that while TNC shapes the immune response during lipopolysaccharide-induced inflammation, this role may be superseded during pneumosepsis caused by a common gram-negative pathogen.
Sigma-Aldrich
Anti-Tenascin Antibody, Chemicon®, from rabbit
Pricing and availability is not currently available.