Merck
  • Helical Carbenium Ion: A Versatile Organic Photoredox Catalyst for Red-Light-Mediated Reactions.
Helical Carbenium Ion: A Versatile Organic Photoredox Catalyst for Red-Light-Mediated Reactions.
Journal of the American Chemical Society (2020-07-01)
Liangyong Mei, José M Veleta, Thomas L Gianetti
ABSTRACT
Red light has the advantages of low energy, less health risks, and high penetration depth through various media. Herein, a helical carbenium ion (N,N'-di-n-propyl-1,13-dimethoxyquinacridinium ( n Pr-DMQA+) tetrafluoroborate) has been used as an organic photoredox catalyst for photoreductions and photooxidations in the presence of red light (λmax = 640 nm). It has catalyzed red-light-mediated dual transition-metal/photo-redox-catalyzed C-H arylation and intermolecular atom-transfer radical addition through oxidative quenching. Moreover, its potential in photooxidation catalysis has also been demonstrated by successful applications in red-light-induced aerobic oxidative hydroxylation of arylboronic acids and benzylic C(sp3)-H oxygenation through reductive quenching. Thus, a versatile organic photoredox catalyst (helical carbenium ion) for red-light-mediated photoredox reactions has been developed.
Sigma-Aldrich
Pr-DMQA[BF4], ≥95%
Pricing and availability is not currently available.