- Rational Design of a Two-Photon Fluorogenic Probe for Visualizing Monoamine Oxidase A Activity in Human Glioma Tissues.
Rational Design of a Two-Photon Fluorogenic Probe for Visualizing Monoamine Oxidase A Activity in Human Glioma Tissues.
Angewandte Chemie (International ed. in English) (2020-02-23)
Haixiao Fang, Hang Zhang, Lin Li, Yun Ni, Riri Shi, Zheng Li, Xuekang Yang, Bo Ma, Chengwu Zhang, Qiong Wu, Changmin Yu, Naidi Yang, Shao Q Yao, Wei Huang
PMID32077158
ABSTRACT
Monoamine oxidases have two functionally distinct but structurally similar isoforms (MAO-A and MAO-B). The ability to differentiate them by using fluorescence detection/imaging technology is of significant biological relevance, but highly challenging with available chemical tools. Herein, we report the first MAO-A-specific two-photon fluorogenic probe (F1), capable of selective imaging of endogenous MAO-A enzymatic activities from a variety of biological samples, including MAO-A-expressing neuronal SY-SY5Y cells, the brain of tumor-bearing mice and human Glioma tissues by using two-photon fluorescence microscopy (TPFM) with minimal cytotoxicity.