Merck
  • TGFβ1, MMPs and cytokines profiles in ocular surface: Possible tear biomarkers for pseudoexfoliation.
TGFβ1, MMPs and cytokines profiles in ocular surface: Possible tear biomarkers for pseudoexfoliation.
PloS one (2021-04-30)
Prity Sahay, Shweta Reddy, Birendra Kumar Prusty, Rahul Modak, Aparna Rao
ABSTRACT
Pseudoexfoliation (PXF) is a unique form of glaucoma characterized by accumulation of exfoliative material in the eyes. Changes in tear profile in disease stages may give us insights into molecular mechanisms involved in causing glaucoma in the eye. All patients were categorized into three main categories; pseudoexfoliation (PXF), pseudoexfoliation glaucoma (PXG) and cataract, which served as control. Cytokines, transforming growth factor β1 (TGFβ1), matrix metalloproteases (MMPs) and fibronectin (FN1) were assessed with multiplex bead assay, enzyme-linked immunosorbent assay (ELISA), gelatin zymography, and immunohistochemistry (IHC) respectively in different ocular tissues such as tears, tenon's capsule, aqueous humor (AH) and serum samples of patients with PXF stages. We found that TGFβ1, MMP-9 and FN1 protein expression were upregulated in tears, tenon's capsule and AH samples in PXG compared to PXF, though the MMP-9 protein activity was downregulated in PXG compared with control or PXF. We have also found that in PXG tears sample the fold change of TGF-α (Transforming Growth Factor-α), MDC (Macrophage Derived Chemokine), IL-8 (Interleukin-8), VEGF (Vascular Endothelial Growth Factor) were significantly downregulated and the levels of GM-CSF (Granulocyte Macrophage Colony Stimulating Factor), IP-10 (Interferon- γ produced protein-10) were significant upregulated. While in AH; IL-6 (Interleukin-6), IL-8, VEGF, IFN-a2 (Interferon- α2), GRO (Growth regulated alpha protein) levels were found lower and IL1a (Interleukin-1α) level was higher in PXG compared to PXF. And in serum; IFN-a2, Eotaxin, GM-CSF, Fractalkine, IL-10 (Interleukin-10), IL1Ra (Interleukin-1 receptor antagonist), IL-7 (Interleukin-7), IL-8, MIP1β (Macrophage Inflammatory Protein-1β), MCP-1 (Monocyte Chemoattractant Protein-1) levels were significantly upregulated and PDGF-AA (Platelet Derived Growth Factor-AA) level was downregulated in the patients with PXG compared to PXF. Altered expression of these molecules in tears may therefore be used as a signal for onset of glaucoma or for identifying eyes at risk of developing glaucoma in PXF.
Sigma-Aldrich
Anti-MMP-9 Antibody, clone 9D4.2, clone 9D4.2, Chemicon®, from mouse
Pricing and availability is not currently available.