Merck
  • Establishment of an iPSC cohort from three unrelated 47-XXY Klinefelter Syndrome patients (KAUSTi007-A, KAUSTi007-B, KAUSTi009-A, KAUSTi009-B, KAUSTi010-A, KAUSTi010-B).
Establishment of an iPSC cohort from three unrelated 47-XXY Klinefelter Syndrome patients (KAUSTi007-A, KAUSTi007-B, KAUSTi009-A, KAUSTi009-B, KAUSTi010-A, KAUSTi010-B).
Stem cell research (2020-10-18)
Maryam Alowaysi, Elisabetta Fiacco, Veronica Astro, Antonio Adamo
ABSTRACT
Klinefelter Syndrome (KS) is caused by the presence of a supernumerary X chromosome. Cytogenetic studies revaled that 80-90% of patients carry a 47-XXY karyotype, while 10-20% of cases are represented by mosaic 46-XY/47-XXY and high-grade aneuploidies 48-XXXY and 48-XXYY. The phenotypic traits of KS are highly variable across individuals and include cognitive dysfunction, metabolic dysregulation, osteoporosis, and cardiovascular diseases. Here, we describe the derivation of multiple 47-XXY iPSC lines from three unrelated KS patients to study the impact of supernumerary X chromosome during early development.
Sigma-Aldrich
Monoclonal Anti-SOX2 antibody produced in mouse, clone 10F10, ascites fluid
Pricing and availability is not currently available.