Merck
  • Identification of capsaicin-sensitive rectal mechanoreceptors activated by rectal distension in mice.
Identification of capsaicin-sensitive rectal mechanoreceptors activated by rectal distension in mice.
Neuroscience (2008-04-09)
N J Spencer, A Kerrin, C A Singer, G W Hennig, W T Gerthoffer, O McDonnell
ABSTRACT
Rodents detect visceral pain in response to noxious levels of rectal distension. However, the mechanoreceptors that innervate the rectum and respond to noxious levels of rectal distension have not been identified. Here, we have identified the mechanoreceptors of capsaicin-sensitive rectal afferents and characterized their properties in response to circumferential stretch of the rectal wall. We have also used the lethal spotted (ls/ls) mouse to determine whether rectal mechanoreceptors that respond to capsaicin and stretch may also develop in an aganglionic rectum that is congenitally devoid of enteric ganglia. In wild type (C57BL/6) mice, graded increases in circumferential stretch applied to isolated rectal segments activated a graded increase in firing of slowly-adapting rectal mechanoreceptors. Identical stimuli applied to the aganglionic rectum of ls/ls mice also activated similar graded increases in firing of stretch-sensitive rectal afferents. In both wild type and aganglionic rectal preparations, focal compression of the serosal surface using von Frey hairs identified mechanosensitive "hot spots," that were associated with brief bursts of action potentials. Spritzing capsaicin (10 microM) selectively onto each identified mechanosensitive hot spot activated an all or none discharge of action potentials in 32 of 56 identified hot spots in wild type mice and 24 of 62 mechanosensitive hot spots in the aganglionic rectum of ls/ls mice. Each single unit activated by both capsaicin and circumferential stretch responded to low mechanical thresholds (1-2 g stretch). No high threshold rectal afferents were ever recorded in response to circumferential stretch. Anterograde labeling from recorded rectal afferents revealed two populations of capsaicin-sensitive mechanoreceptor that responded to stretch: one population terminated within myenteric ganglia, the other within the circular and longitudinal smooth muscle layers. In the aganglionic rectum of ls/ls mice, only the i.m. mechanoreceptors were identified. Both myenteric and i.m. mechanoreceptors could be identified by their immunoreactivity to the anti-TRPV1 antibody and the vesicular glutamate transporter, Vglut2. Myenteric mechanoreceptors had a unique morphology, consisting of smooth bulbous nodules that ramified within myenteric ganglia. In summary, the rectum of wild type mice is innervated by at least two populations of capsaicin-sensitive rectal mechanoreceptor, both of which respond to low mechanical thresholds within the innocuous range. These findings suggest that the visceral pain pathway activated by rectal distension is likely to involve low threshold rectal mechanoreceptors that are activated within the normal physiological range.
Sigma-Aldrich
Anti-Capsaicin Receptor (Ab-1) (824-838) Rabbit pAb, liquid, Calbiochem®