Merck
  • The Expression of BTS-2 Enhances Cell Growth and Invasiveness in Renal Cell Carcinoma.
The Expression of BTS-2 Enhances Cell Growth and Invasiveness in Renal Cell Carcinoma.
Anticancer research (2017-05-30)
Quoc Thang Pham, Naohide Oue, Yuji Yamamoto, Yoshinori Shigematsu, Yohei Sekino, Naoya Sakamoto, Kazuhiro Sentani, Naohiro Uraoka, Mamata Tiwari, Wataru Yasui
ABSTRACT
Renal cell carcinoma (RCC) is one of the most common types of cancer in developed countries. Bone marrow stromal cell antigen 2 (BST2) gene, which encodes BST2 transmembrane glycoprotein, is overexpressed in several cancer types. In the present study, we analyzed the expression and function of BST2 in RCC. BST2 expression was analyzed by immunohistochemistry in 123 RCC cases. RNA interference was used to inhibit BST2 expression in a RCC cell line. Immunohistochemical analysis showed that 32% of the 123 RCC cases were positive for BST2. BST2 expression was positively associated with tumour stage. Furthermore, BST2 expression was an independent predictor of survival in patients with RCC. BST2 siRNA-transfected Caki-1 cells displayed significantly reduced cell growth and invasive activity relative to negative control siRNA-transfected cells. These results suggest that BST2 plays an important role in the progression of RCC. Because BST2 is expressed on the cell membrane, BST2 is a good therapeutic target for RCC.
Sigma-Aldrich
Anti-BST2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.