Merck
  • Genomic RNA Elements Drive Phase Separation of the SARS-CoV-2 Nucleocapsid.
Genomic RNA Elements Drive Phase Separation of the SARS-CoV-2 Nucleocapsid.
Molecular cell (2020-12-09)
Christiane Iserman, Christine A Roden, Mark A Boerneke, Rachel S G Sealfon, Grace A McLaughlin, Irwin Jungreis, Ethan J Fritch, Yixuan J Hou, Joanne Ekena, Chase A Weidmann, Chandra L Theesfeld, Manolis Kellis, Olga G Troyanskaya, Ralph S Baric, Timothy P Sheahan, Kevin M Weeks, Amy S Gladfelter
ABSTRACT
We report that the SARS-CoV-2 nucleocapsid protein (N-protein) undergoes liquid-liquid phase separation (LLPS) with viral RNA. N-protein condenses with specific RNA genomic elements under physiological buffer conditions and condensation is enhanced at human body temperatures (33°C and 37°C) and reduced at room temperature (22°C). RNA sequence and structure in specific genomic regions regulate N-protein condensation while other genomic regions promote condensate dissolution, potentially preventing aggregation of the large genome. At low concentrations, N-protein preferentially crosslinks to specific regions characterized by single-stranded RNA flanked by structured elements and these features specify the location, number, and strength of N-protein binding sites (valency). Liquid-like N-protein condensates form in mammalian cells in a concentration-dependent manner and can be altered by small molecules. Condensation of N-protein is RNA sequence and structure specific, sensitive to human body temperature, and manipulatable with small molecules, and therefore presents a screenable process for identifying antiviral compounds effective against SARS-CoV-2.
Sigma-Aldrich
Atto 488 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
1,6-Hexanediol, 99%
Sigma-Aldrich
(R)-(+)-α-Lipoic acid, ≥98.0% (HPLC)