Merck
  • MYC Regulates PHF8, Which Promotes the Progression of Gastric Cancer by Suppressing miR-22-3p.
MYC Regulates PHF8, Which Promotes the Progression of Gastric Cancer by Suppressing miR-22-3p.
Technology in cancer research & treatment (2020-10-29)
Ming-Zhi Cai, Shao-Yan Wen, Xue-Jun Wang, Yong Liu, Han Liang
ABSTRACT
Plant homeodomain finger protein 8 (PHF8) has been reported to participate in cancer development and metastasis of various types of tumors. However, little is known about the functional mechanism of PHF8 in gastric cancer (GC). This study aimed to explore the PHF8 expression pattern and function, and the role of the MYC/miRNA/PHF8 axis in GC. PHF8 expression was upregulated in GC tissues and cells as measured using quantitative reverse transcription polymerase chain reaction and western blotting. PHF8 knockdown suppressed the proliferation, migration, and invasion of GC cells, as determined using the CCK-8 assay and Transwell assay. MicroRNA-22-3p targeted PHF8, as verified by a dual-luciferase reporter assay. MYC upregulated the protein expression of PHF8 but had no effect on PHF8 mRNA expression. MYC regulates PHF8 by affecting the stability of miR-22-3p. We identified a novel MYC/miR-22-3p/PHF8 regulatory axis in GC. Therefore, PHF8 may provide a new therapeutic target for patients with GC.
Sigma-Aldrich
MISSION® esiRNA, targeting human PHF8