Merck
  • fMRI indicates cortical activation through TRPV1 modulation during acute gouty attacks.
fMRI indicates cortical activation through TRPV1 modulation during acute gouty attacks.
Scientific reports (2019-08-28)
Chiao-Chi Chen, Chen Chang, Yi-Hua Hsu, Yi-Jen Peng, Herng-Sheng Lee, Guo-Shu Huang
ABSTRACT
Gout is one of the most painful disease conditions. The central mechanism of pain processing in this condition remains elusive. Cerebral blood volume (CBV) responses are faithful correlates of brain activity changes; the application of CBV-weighted functional magnetic resonance imaging (fMRI) may shed light on the issue of interest. Transient receptor potential vanilloid 1 (TRPV1) is a critical ion channel expressed both peripherally in nociceptors and centrally in the brain. Whether TRPV1 plays a critical role in gout pain was also explored. Results showed that, in rats with gouty arthritis, noxious stimulation induced CBV increases in the primary somatosensory cortex and thalamus. These increases were correlated with up-regulated TRPV1 protein expression and pain behavior. Selective blockage of central TRPV1 channel activity by intrathecal administration of AMG9810 reversed the induced pain, and abolished the induced CBV increase in thalamocortical regions. The findings support that TRPV1 activation in the central pain pathway is crucial to the augmentation of pain in gouty conditions. This new information supports the development of TRPV1-based drugs for treating gout pain, while fMRI can be useful for repeated evaluation of brain activity changes induced by gout.
Sigma-Aldrich
Uric acid, ≥99%, crystalline
Sigma-Aldrich
Anti-Capsaicin Receptor (Ab-1) (824-838) Rabbit pAb, liquid, Calbiochem®
Supelco
1,25-Dihydroxyvitamin D3-13C3 (25,26,27-13C3) solution, 5 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®