Merck
  • Effects of injectable calcitriol on mineral metabolism and postpartum health and performance in dairy cows.
Effects of injectable calcitriol on mineral metabolism and postpartum health and performance in dairy cows.
Journal of dairy science (2020-11-10)
A Vieira-Neto, G Negro, R Zimpel, M Poindexter, F Lopes, W W Thatcher, C D Nelson, J E P Santos
ABSTRACT
Objectives were to determine the effects of an injectable formulation of calcitriol on Ca concentration, risk of clinical diseases, and performance in dairy cows. Cows were blocked by lactation number (1 vs. >1) and calving sequence and, within block, assigned randomly within 6 h of calving to receive subcutaneously vehicle only (CON, n = 450) or 200 (CAL200, n = 450) or 300 μg of 1α,25-dihydroxyvitamin D3 (CAL300, n = 450). Cows were fed the same acidogenic diet prepartum. Blood was sampled before treatment administration and again during the first 11 d postpartum and analyzed for concentrations of ionized Ca (iCa), total Ca (tCa), Mg (tMg), and P (tP), β-hydroxybutyrate, carboxylated osteocalcin (cOC), and undercarboxylated osteocalcin (uOC). Cows were evaluated for diseases in the first 60 d postpartum. Reproduction and survival were monitored for the first 300 d postpartum. Calcitriol increased concentration of blood iCa (CON = 1.12 vs. CAL200 = 1.23 vs. CAL300 = 1.27 mM), plasma tCa (CON = 2.29 vs. CAL200 = 2.44 vs. CAL300 = 2.46 mM), and plasma tP (CON = 1.72 vs. CAL200 = 2.21 vs. CAL300 = 2.28 mM), and differences were observed during the first 5 d postpartum for iCa and tCa, and the first 7 d postpartum for tP. Concentrations of tMg were lower in calcitriol-treated cows than in CON cows (CON = 0.81 vs. CAL200 = 0.78 vs. CAL300 = 0.75 mM), and differences were observed during the first 5 d postpartum. Calcitriol increased plasma concentrations of cOC (CON = 14.5 vs. CAL200 = 23.0 vs. CAL300 = 19.8 ng/mL) and uOC (CON = 1.6 vs. CAL200 = 3.4 vs. CAL300 = 2.6 ng/mL). Prevalence of subclinical hypocalcemia was less in calcitriol-treated cows (CON = 19.0 vs. CAL200 = 4.7 vs. CAL300 = 9.3%); however, benefits on health were only observed in overconditioned cows (n = 270/1,350). Calcitriol reduced incidence of retained placenta (CON = 14.3 vs. CAL200 = 5.1 vs. CAL300 = 5.9%), puerperal metritis (CON = 12.7 vs. CAL200 = 6.1 vs. CAL300 = 2.5%), and morbidity (CON = 72.1 vs. CAL200 = 57.4 vs. CAL300 = 56.9%) in cows with BCS greater than 3.50, but no benefit on health was observed in cows with BCS equal to or less than 3.50 at parturition. Milk yield did not differ among treatments. Pregnancy at first AI did not differ, but pregnancy rate after the first AI was slower for calcitriol-treated cows because of reduced insemination rate and pregnancy per AI. We found that CAL200 reduced death but increased culling in cows without calving problems. Collectively, results indicate that treatment with calcitriol at parturition was effective in improving concentrations of iCa, tCa, and tP, which reduced the risk of hypocalcemia. Pregnancy rate was reduced by calcitriol treatment, and benefits on health performance were limited to overconditioned cows. Thus, treatment of all cows is not supported, and proper identification of cohorts of cows that benefit from postpartum interventions that increase blood calcitriol or calcium is needed.
Sigma-Aldrich
2-Hydroxy-4-(methylthio)butyric acid calcium salt, ≥99.0% (NT)
Pricing and availability is not currently available.