Merck
  • Analysis of the functional group composition of the spruce and birch phloem lignin.
Analysis of the functional group composition of the spruce and birch phloem lignin.
International journal of biological macromolecules (2020-11-05)
Anna V Faleva, Artem V Belesov, Aleksandr Yu Kozhevnikov, Danil I Falev, Dmitry G Chukhchin, Evgeniy V Novozhilov
ABSTRACT
In this article, the functional group composition of the spruce (Pícea ábies) and birch (Bétula péndula) phloem lignin is described. The features of the chemical structure were studied by analyzing dioxane lignin using the elemental analysis, UV-Vis, FT-IR, and 1D NMR spectroscopy. For comparison, samples of xylem dioxane lignin isolated from the corresponding wood species were also analyzed. FT-IR spectroscopy data suggest that the lignins of birch phloem and xylem are similar in chemical structure. However, there are differences in absorption bands in the spectra of spruce dioxane lignin, which indicate the opposite. Quantitative analysis of the functional group composition was performed using 13C and 31P NMR data. It was found that free phenolic hydroxyl groups of catechol and p-hydroxyphenyl types are dominated in the composition of spruce phloem lignin. Birch phloem lignin has a qualitative and quantitative composition of functional groups characteristic of hardwood lignins. However, the content of G-units is greater than S-units, in contrast to the birch xylem lignin, where S-units predominate. The revealed differences are relevant from the point of view of plant physiology. The practical significance of the study is connected with understanding the reactivity of lignins when considering the chemical processing of tree bark.
Sigma-Aldrich
N-Hydroxy-5-norbornene-2,3-dicarboxylic acid imide, 97%
Sigma-Aldrich
2-Chloro-4,4,5,5-tetramethyl-1,3,2-dioxaphospholane, 95%