Merck
  • CD14+ monocytes and CD163+ macrophages correlate with the severity of liver fibrosis in patients with chronic hepatitis C.
CD14+ monocytes and CD163+ macrophages correlate with the severity of liver fibrosis in patients with chronic hepatitis C.
Experimental and therapeutic medicine (2020-11-06)
Su-Xian Zhao, Wen-Cong Li, Na Fu, Ling-Bo Kong, Qing-Shan Zhang, Fang Han, Wei-Guang Ren, Po Cui, Jing-Hua Du, Bao-Yu Wang, Yu-Guo Zhang, Rong-Qi Wang, Li Kong, Yue-Min Nan
ABSTRACT
Hepatic fibrosis is a crucial pathological process involved in the development of chronic hepatitis C (CHC) and may progress to liver cirrhosis and hepatocellular carcinoma. Activated peripheral blood monocytes and intrahepatic macrophages further promote hepatic fibrogenesis by releasing proinflammatory and profibrogenic cytokines. The present study aimed to investigate the role of peripheral CD14+ monocytes and intrahepatic CD163+ macrophages in hepatitis C virus (HCV)-associated liver fibrosis and clarify whether serum soluble CD163 (sCD163) may serve as a fibrosis marker in patients with CHC. A total of 87 patients with CHC and 20 healthy controls were recruited. Serum sCD163 levels were measured by ELISA. Frequencies of peripheral CD14+ monocytes and inflammatory cytokines expressed by CD14+ monocytes were analyzed by flow cytometry. The degree of fibrosis in human liver biopsies was graded using the Metavir scoring system and patients were stratified into two groups based on those results (F<2 vs. F≥2). Hepatic expression of CD163 was examined by immunohistochemical staining. The diagnostic values of sCD163, aspartate aminotransferase to platelet ratio index (APRI), fibrosis 4 score (FIB-4) and the aspartate aminotransferase to alanine aminotransferase ratio (AAR) in significant fibrosis (F≥2) were evaluated and compared using receiver operating characteristic (ROC) curves. The results indicated that the serum sCD163 levels and the frequency of CD14+ monocytes were significantly higher in the patients than that in the controls and positively correlated with liver fibrosis. The level of serum sCD163 was consistent with hepatic CD163 expression in the liver sections from patients. The frequencies of interleukin (IL)-8- and tumor necrosis factor-α-expressing monocytes were increased and that of IL-10-expressing monocytes was decreased in the patients. The area under the ROC curve (AUROC) for sCD163, APRI, FIB-4 and AAR was 0.876, 0.785, 0.825 and 0.488, respectively, and the AUROC for sCD163 was significantly higher than those for APRI and AAR. In conclusion, sCD163 may serve as a novel marker for assessing the degree of liver fibrosis in HCV-infected patients.
Sigma-Aldrich
Human CD163 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Pricing and availability is not currently available.