Merck
  • Characterization of rare earth elements present in coal ash by sequential extraction.
Characterization of rare earth elements present in coal ash by sequential extraction.
Journal of hazardous materials (2020-12-02)
Sungyoon Park, Minsoo Kim, Yejee Lim, Jimin Yu, Siyu Chen, Sang Woon Woo, Sunho Yoon, Sungjun Bae, Han S Kim
ABSTRACT
Although it has recently been reported that notable amounts of rare earth elements (REEs) are present in the residual coal ash, little is currently known regarding the association of these elements with the coal ash matrix, thereby limiting the potential for extraction of REEs from coal ash. In this study, we analyzed the binding characteristics of REEs within coal ash via sequential extraction and examined REE recovery during a coal ash recycling process. Major components of coal ash were found to be mineral oxides, mainly composed of Si, Fe, Al, and Ca, and residual carbons. Bottom and fly ashes were found to contain 185.8 mg/kg and 179.2 mg/kg of REEs, respectively. Tessier sequential extraction confirmed that 85 % of REEs are included in the residual fraction of both bottom and fly ashes. Furthermore, BCR sequential extraction revealed that 60-70 % of REEs are contained within the residual fraction, thereby indicating that REEs are strongly bound in both bottom and fly ashes and the use of very strong acids is required for the thorough extraction of REEs from coal ash. Additionally, it was found that 46.3 % of REEs can be recovered from the wastewaters produced during the process of coal ash-derived zeolite synthesis.
Sigma-Aldrich
Hydroxylamine hydrochloride, ≥97.0%, suitable for determination of toxic metals
Sigma-Aldrich
Sodium acetate, JIS special grade, ≥98.5%