Merck
  • Peptide-protein coassembling matrices as a biomimetic 3D model of ovarian cancer.
Peptide-protein coassembling matrices as a biomimetic 3D model of ovarian cancer.
Science advances (2020-10-04)
Clara Louise Hedegaard, Carlos Redondo-Gómez, Bee Yi Tan, Kee Woei Ng, Daniela Loessner, Alvaro Mata
ABSTRACT
Bioengineered three-dimensional (3D) matrices expand our experimental repertoire to study tumor growth and progression in a biologically relevant, yet controlled, manner. Here, we used peptide amphiphiles (PAs) to coassemble with and organize extracellular matrix (ECM) proteins producing tunable 3D models of the tumor microenvironment. The matrix was designed to mimic physical and biomolecular features of tumors present in patients. We included specific epitopes, PA nanofibers, and ECM macromolecules for the 3D culture of human ovarian cancer, endothelial, and mesenchymal stem cells. The multicellular constructs supported the formation of tumor spheroids with extensive F-actin networks surrounding the spheroids, enabling cell-cell communication, and comparative cell-matrix interactions and encapsulation response to those observed in Matrigel. We conducted a proof-of-concept study with clinically used chemotherapeutics to validate the functionality of the multicellular constructs. Our study demonstrates that peptide-protein coassembling matrices serve as a defined model of the multicellular tumor microenvironment of primary ovarian tumors.
Sigma-Aldrich
Hematoxylin Solution, Gill No. 1
Pricing and availability is not currently available.
Sigma-Aldrich
Debrisoquine sulfate
Pricing and availability is not currently available.