Merck
  • Micellar electrokinetic chromatography as efficient alternative for the multiresidue determination of seven neonicotinoids and 6-chloronicotinic acid in environmental samples.
Micellar electrokinetic chromatography as efficient alternative for the multiresidue determination of seven neonicotinoids and 6-chloronicotinic acid in environmental samples.
Analytical and bioanalytical chemistry (2019-12-12)
Laura Carbonell-Rozas, Francisco J Lara, Monsalud Del Olmo Iruela, Ana M García-Campaña
ABSTRACT
A simple, sensitive, and efficient method has been developed for the determination of the seven neonicotinoid insecticides commercially available (imidacloprid, thiacloprid, clothianidin, thiamethoxam, acetamiprid, nitenpyram, and dinotefuran) and the main metabolite 6-chloronicotinic acid. Micellar electrokinetic chromatography (MEKC) mode was applied, using 48.5 cm of total length capillary (50 μm i.d.) with an extended light-path capillary (150 μm). The running electrolyte consisted of 25 mM sodium tetraborate buffer (pH 9.2) containing 120 mM of sodium dodecyl sulfate and 15% of methanol (v/v). A voltage of 27 kV and a temperature of 25 °C were applied. Samples dissolved in deionized water were hydrodynamically injected at 50 mbar for 12 s, achieving the analysis in less than 12 min. Diode array detection (DAD) was performed at 220, 254, and 270 nm, depending on the analyte. Two different methodologies as sample treatments were developed; for water samples, solid-phase extraction was checked using different cartridges (C18, Oasis® HLB, Oasis® HLB Prime, and Strata-X), being the best option Oasis® HLB for preconcentration and cleanup. In the case of soil samples, a simple solid-liquid extraction was applied using a mixture of 1:3 (v/v) acetonitrile/dichloromethane. Satisfactory linearity, trueness, and precision were achieved, with detection limits in the range of 0.1-0.4 μg L-1 for river water and 1.0-2.9 μg kg-1 for soil samples. Recoveries in the range of 80-107% for all of the assayed neonicotinoids in water samples of different origin and 73-92% for soil samples were achieved.
Product Description
Supelco
Nitenpyram, PESTANAL®, analytical standard
Pricing and availability is not currently available.