Merck
  • Ex vivo editing of human hematopoietic stem cells for erythroid expression of therapeutic proteins.
Ex vivo editing of human hematopoietic stem cells for erythroid expression of therapeutic proteins.
Nature communications (2020-07-31)
Giulia Pavani, Marine Laurent, Anna Fabiano, Erika Cantelli, Aboud Sakkal, Guillaume Corre, Peter J Lenting, Jean-Paul Concordet, Magali Toueille, Annarita Miccio, Mario Amendola
ABSTRACT
Targeted genome editing has a great therapeutic potential to treat disorders that require protein replacement therapy. To develop a platform independent of specific patient mutations, therapeutic transgenes can be inserted in a safe and highly transcribed locus to maximize protein expression. Here, we describe an ex vivo editing approach to achieve efficient gene targeting in human hematopoietic stem/progenitor cells (HSPCs) and robust expression of clinically relevant proteins by the erythroid lineage. Using CRISPR-Cas9, we integrate different transgenes under the transcriptional control of the endogenous α-globin promoter, recapitulating its high and erythroid-specific expression. Erythroblasts derived from targeted HSPCs secrete different therapeutic proteins, which retain enzymatic activity and cross-correct patients' cells. Moreover, modified HSPCs maintain long-term repopulation and multilineage differentiation potential in transplanted mice. Overall, we establish a safe and versatile CRISPR-Cas9-based HSPC platform for different therapeutic applications, including hemophilia and inherited metabolic disorders.
Sigma-Aldrich
4-Methylumbelliferyl phosphate disodium salt, phosphatase substrate
Pricing and availability is not currently available.