Merck
  • Development of natural cellulase inhibitor mediated intensified biological pretreatment technology using Pleurotus florida for maximum recovery of cellulose from paddy straw under solid state condition.
Development of natural cellulase inhibitor mediated intensified biological pretreatment technology using Pleurotus florida for maximum recovery of cellulose from paddy straw under solid state condition.
Bioresource technology (2017-08-07)
Manickam Naresh Kumar, Rajarathinam Ravikumar, Senniyappan Thenmozhi, Muthuvelu Kirupa Sankar
ABSTRACT
Inhibitor mediated intensified bio-pretreatment (IMBP) technology using natural cellulase inhibitor (NCI) for maximum cellulose recovery from paddy straw was studied. Pretreatment was carried out under solid state condition. Supplementation of 8% NCI in pretreatment medium improves cellulose recovery and delignification by 1.2 and 1.5-fold respectively, compared to conventional bio-pretreatment due to inhibition of 61% of cellulase activity in IMBP. Further increase in NCI concentration showed negative effect on Pleurotus florida growth and suppress the laccase productivity by 1.1-fold. Laccase activity in IMBP was found to be 2.0U/mL on 19thday, which is higher than (1.5U/mL) conventional bio-pretreatment. Physico-chemical modifications in paddy straw before and after pretreatment were analysed by SEM, ATR-FTIR, XRD and TGA. According to these findings, the IMBP technology can be a viable eco-friendly technology for sustainable production of bioethanol with maximum cellulose recovery.
Sigma-Aldrich
Syringaldazine, 98%
Pricing and availability is not currently available.