Merck
  • Hic-5 promotes the hypertrophic scar myofibroblast phenotype by regulating the TGF-beta1 autocrine loop.
Hic-5 promotes the hypertrophic scar myofibroblast phenotype by regulating the TGF-beta1 autocrine loop.
The Journal of investigative dermatology (2008-04-11)
Ganary Dabiri, David A Tumbarello, Christopher E Turner, Livingston Van de Water
ABSTRACT
Following severe traumatic or thermal injury to the dermis, hypertrophic scars (HTSs) often develop in humans. These scar fibroblasts (hypertrophic scar fibroblasts (HTSFs)) retain the myofibroblast phenotype persistently, rather than transiently as in acute wounds. These pathogenic myofibroblasts constitutively express smooth-muscle cell alpha-actin (SMAA), deposit an excessive amount of extracellular matrix (ECM) proteins, are highly contractile, and stably display large focal adhesions. Increasing evidence supports a mechanism in which autocrine production and activation of transforming growth factor-beta1 (TGF-beta1) are required to maintain the pathogenic myofibroblast phenotype. We recently reported that Hic-5, a focal adhesion protein that is upregulated by TGF-beta1, is expressed persistently in HTSF compared to normal adult fibroblasts (NADFs). We now find that Hic-5 is an important regulator of the constitutive myofibroblast phenotype in HTSFs. Silencing the expression of Hic-5 in HTSFs with specific siRNAs dramatically reduces TGF-beta1 production, decreases the generation of supermature focal adhesions reduces expression of SMAA and decreases collagen contraction and ECM synthesis. Our findings demonstrate that Hic-5 is an essential component of the mechanism regulating autocrine production of TGF-beta1 and the resulting pathogenic myofibroblast phenotype.
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered