Merck
  • Cross-linked cartilage acellular matrix film decreases postsurgical peritendinous adhesions.
Cross-linked cartilage acellular matrix film decreases postsurgical peritendinous adhesions.
Artificial organs (2019-10-30)
Do Young Park, Hee-Woong Yun, Sumin Lim, Minh-Dung Truong, Xiang Yun Yin, Jinho Park, Byeong Kook Kim, Dong Il Shin, Xue Guang Li, Jun Young Chung, Moon Suk Kim, Byoung-Hyun Min
ABSTRACT
Cartilage extracellular matrix contains antiadhesive and antiangiogenic molecules such as chondromodulin-1, thrombospondin-1, and endostatin. We have aimed to develop a cross-linked cartilage acellular matrix (CAM) barrier for peritendinous adhesion prevention. CAM film was fabricated using decellularized porcine cartilage tissue powder and chemical cross-linking. Biochemical analysis of the film showed retention of collagen and glycosaminoglycans after the fabrication process. Physical characterization of the film showed denser collagen microstructure, increased water contact angle, and higher tensile strength after cross-linking. The degradation time in vivo was 14 d after cross-linking. The film extract and film surface showed similar cell proliferation, while inhibiting cell migration and cell adhesion compared to standard media and culture plate, respectively. Application of the film after repair resulted in similar tendon healing and significantly less peritendinous adhesions in a rabbit Achilles tendon injury model compared to repair only group, demonstrated by histology, ultrasonography, and biomechanical testing. In conclusion, the current study developed a CAM film having biological properties of antiadhesion, together with biomechanical properties and degradation profile suitable for prevention of peritendinous adhesions.
Sigma-Aldrich
QCM Chemotaxis Cell Migration Assay, 24-well (8 µm), colorimetric, The QCM 24-well Migration Assay is ideal for the study of chemotaxis cell migration. The assay uses a 24-well plate with an 8 micron pore size, with colorimetric detection.
Pricing and availability is not currently available.